gRNA Design Details
- Target DNA Sequence
- GCGGAAACTTCGAGGCATGA
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 19603240 - 19603262 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Clasrp
- Entrez Gene ID
-
53609
- Gene Name
- CLK4-associating serine/arginine rich protein
- Gene Aliases
-
Clasp, Sfrs16, Srsf16, Swap2
- Chromosome Location
-
Chr.7: 19581038 - 19604517 on GRCm38
- UniGene ID
- Mm.20913
- Species
-
Mouse