gRNA Design Details
- Target DNA Sequence
- AAAGCGGTCAATCATGTTGT
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 19594966 - 19594988 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.19/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Clasrp
- Entrez Gene ID
-
53609
- Gene Name
- CLK4-associating serine/arginine rich protein
- Gene Aliases
-
Clasp, Sfrs16, Srsf16, Swap2
- Chromosome Location
-
Chr.7: 19581038 - 19604517 on GRCm38
- UniGene ID
- Mm.20913
- Species
-
Mouse