gRNA Design Details
- Target DNA Sequence
- TGAAGATCCTCGGGAACAGG
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 171775023 - 171775045 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.81/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Slamf1
- Entrez Gene ID
-
27218
- Gene Name
- signaling lymphocytic activation molecule family member 1
- Gene Aliases
-
4933415F16, AA177906, CD150, CDw150, ESTM51, IPO-3, Slam
- Chromosome Location
-
Chr.1: 171767132 - 171801184 on GRCm38
- UniGene ID
- Mm.103648
- Species
-
Mouse