gRNA Design Details
- Target DNA Sequence
- AAGTCGTGTGGTCCCTGATC
- PAM Sequence
- GGG
- Target locus
-
Chr.9: 64020958 - 64020980 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.59/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Smad6
- Entrez Gene ID
-
17130
- Gene Name
- SMAD family member 6
- Gene Aliases
-
Madh6, b2b390Clo
- Chromosome Location
-
Chr.9: 63953076 - 64022085 on GRCm38
- UniGene ID
- Mm.325757
- Species
-
Mouse