gRNA Design Details
- Target DNA Sequence
- ATCCTGATGAGATGCGCAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 36000245 - 36000267 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.02/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Kpna1
- Entrez Gene ID
-
16646
- Gene Name
- karyopherin (importin) alpha 1
- Gene Aliases
-
AW494490, IPOA5, NPI1, Rch2, mSRP1
- Chromosome Location
-
Chr.16: 35983363 - 36037136 on GRCm38
- UniGene ID
- Mm.6952
- Species
-
Mouse