gRNA Design Details
- Target DNA Sequence
- TATGAATGTGTCTCCTCGAT
- PAM Sequence
- AGG
- Target locus
-
Chr.13: 89042486 - 89042508 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Edil3
- Entrez Gene ID
-
13612
- Gene Name
- EGF-like repeats and discoidin I-like domains 3
- Gene Aliases
-
Del-1, Del1
- Chromosome Location
-
Chr.13: 88821472 - 89323225 on GRCm38
- UniGene ID
- Mm.125580
- Species
-
Mouse