gRNA Design Details
- Target DNA Sequence
- TCGTGGATTCTGACAAGAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 115857059 - 115857081 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.77/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mknk1
- Entrez Gene ID
-
17346
- Gene Name
- MAP kinase-interacting serine/threonine kinase 1
- Gene Aliases
-
2410048M24Rik, Mnk1
- Chromosome Location
-
Chr.4: 115838984 - 115879256 on GRCm38
- UniGene ID
- Mm.209327
- Species
-
Mouse