gRNA Design Details
- Target DNA Sequence
- AGAGACGCTACAATTCGCCC
- PAM Sequence
- CGG
- Target locus
-
Chr.6: 126102194 - 126102216 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ntf3
- Entrez Gene ID
-
18205
- Gene Name
- neurotrophin 3
- Gene Aliases
-
AI316846, AI835689, Nt3, Ntf-3
- Chromosome Location
-
Chr.6: 126101412 - 126166791 on GRCm38
- UniGene ID
- Mm.267570
- Species
-
Mouse