gRNA Design Details
- Target DNA Sequence
- GACCATCTTCGGGTGAACTG
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 32436216 - 32436238 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
93.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pik3ca
- Entrez Gene ID
-
18706
- Gene Name
- phosphatidylinositol 3-kinase, catalytic, alpha polypeptide
- Gene Aliases
-
6330412C24Rik, caPI3K, p110, p110alpha
- Chromosome Location
-
Chr.3: 32397055 - 32468486 on GRCm38
- UniGene ID
- Mm.260521
- Species
-
Mouse