gRNA Design Details
- Target DNA Sequence
- GACTTTAGAATGCCTCCGTG
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 179198921 - 179198943 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
71.95/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PIK3CA
- Entrez Gene ID
-
5290
- Gene Name
- phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha
- Gene Aliases
-
CLOVE, CWS5, MCAP, MCM, MCMTC, PI3K, PI3K-alpha, p110-alpha
- Chromosome Location
-
Chr.3: 179148114 - 179240093 on GRCh38
- UniGene ID
- Hs.553498
- Species
-
Human