gRNA Design Details
- Target DNA Sequence
- ACATTCCCCGTCAAATTTAC
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 50930594 - 50930616 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmeff2
- Entrez Gene ID
-
56363
- Gene Name
- transmembrane protein with EGF-like and two follistatin-like domains 2
- Gene Aliases
-
4832418D20Rik, 7630402F16Rik, TR-2
- Chromosome Location
-
Chr.1: 50909962 - 51187270 on GRCm38
- UniGene ID
- Mm.245154
- Species
-
Mouse