gRNA Design Details
- Target DNA Sequence
- AAGCAGCGAGTTTTACCGGG
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 50928046 - 50928068 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.4/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmeff2
- Entrez Gene ID
-
56363
- Gene Name
- transmembrane protein with EGF-like and two follistatin-like domains 2
- Gene Aliases
-
4832418D20Rik, 7630402F16Rik, TR-2
- Chromosome Location
-
Chr.1: 50909962 - 51187270 on GRCm38
- UniGene ID
- Mm.245154
- Species
-
Mouse