gRNA Design Details
- Target DNA Sequence
- CTGCTCGCACTGTTGAGTCT
- PAM Sequence
- CGG
- Target locus
-
Chr.7: 141192197 - 141192219 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
83.03/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Hras
- Entrez Gene ID
-
15461
- Gene Name
- Harvey rat sarcoma virus oncogene
- Gene Aliases
-
H-ras, Ha-ras, Harvey-ras, Hras-1, Hras1, Kras2, c-H-ras, c-Ha-ras, c-rasHa, ras
- Chromosome Location
-
Chr.7: 141189934 - 141194004 on GRCm38
- UniGene ID
- Mm.334313
- Species
-
Mouse