gRNA Design Details
- Target DNA Sequence
- GAGCCTGTCCTCATTCGAAA
- PAM Sequence
- AGG
- Target locus
-
Chr.18: 43728173 - 43728195 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spink1
- Entrez Gene ID
-
20730
- Gene Name
- serine peptidase inhibitor, Kazal type 1
- Gene Aliases
-
Spink3, p12
- Chromosome Location
-
Chr.18: 43728069 - 43737237 on GRCm38
- UniGene ID
- Mm.272
- Species
-
Mouse