gRNA Design Details
- Target DNA Sequence
- TAGTTGCCATGATGCAGTGG
- PAM Sequence
- CGG
- Target locus
-
Chr.18: 43733606 - 43733628 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.99/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spink1
- Entrez Gene ID
-
20730
- Gene Name
- serine peptidase inhibitor, Kazal type 1
- Gene Aliases
-
Spink3, p12
- Chromosome Location
-
Chr.18: 43728069 - 43737237 on GRCm38
- UniGene ID
- Mm.272
- Species
-
Mouse