gRNA Design Details
- Target DNA Sequence
- ACCTCTTCGTCCAGAAATTG
- PAM Sequence
- GGG
- Target locus
-
Chr.1: 75211387 - 75211409 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk16
- Entrez Gene ID
-
20872
- Gene Name
- serine/threonine kinase 16
- Gene Aliases
-
EDPK, Krct, MPSK, PKL12, TSF-1
- Chromosome Location
-
Chr.1: 75210829 - 75215606 on GRCm38
- UniGene ID
- Mm.1155
- Species
-
Mouse