gRNA Design Details
- Target DNA Sequence
- GATGCATCTCTGCCTCTCGT
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 75212024 - 75212046 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.13/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk16
- Entrez Gene ID
-
20872
- Gene Name
- serine/threonine kinase 16
- Gene Aliases
-
EDPK, Krct, MPSK, PKL12, TSF-1
- Chromosome Location
-
Chr.1: 75210829 - 75215606 on GRCm38
- UniGene ID
- Mm.1155
- Species
-
Mouse