gRNA Design Details

Target DNA Sequence
AGGTGGTAAGCTCAAGCCGG
PAM Sequence
AGG
Target locus
Chr.4: 41739377 - 41739399 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 93.77/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Sigmar1
Entrez Gene ID
18391
Gene Name
sigma non-opioid intracellular receptor 1
Gene Aliases
Oprs1, sigma1R
Chromosome Location
Chr.4: 41738493 - 41741359 on GRCm38
UniGene ID
Mm.425181
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001286538.1 NP_001273467.1 4
NM_001286539.1 NP_001273468.1 3
NM_001286540.1 NP_001273469.1 4
NM_001286541.1 NP_001273470.1 3
NM_001286542.1 NP_001273471.1 3
NM_001286551.1 NP_001273480.1 4
NM_001286605.1 NP_001273534.1 4
NM_011014.3 NP_035144.1 4