gRNA Design Details

Target DNA Sequence
GTACTTGGTAATGGAACTAA
PAM Sequence
TGG
Target locus
Chr.11: 49869172 - 49869194 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 85.08/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Mapk9
Entrez Gene ID
26420
Gene Name
mitogen-activated protein kinase 9
Gene Aliases
AI851083, JNK2, Prkm9, p54aSAPK
Chromosome Location
Chr.11: 49846751 - 49886421 on GRCm38
UniGene ID
Mm.68933
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001163671.1 NP_001157143.1 5
NM_001163672.1 NP_001157144.1 5
NM_016961.3 NP_058657.1 5
NM_207692.2 NP_997575.2 5