gRNA Design Details

Target DNA Sequence
GGTCCATCAACAGCTGCTCC
PAM Sequence
AGG
Target locus
Chr.3: 37125744 - 37125766 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 81.9/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Il2
Entrez Gene ID
16183
Gene Name
interleukin 2
Gene Aliases
Il-2
Chromosome Location
Chr.3: 37120713 - 37125954 on GRCm38
UniGene ID
Mm.14190
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_008366.3 NP_032392.1 1