gRNA Design Details
- Target DNA Sequence
- GGTCCATCAACAGCTGCTCC
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 37125744 - 37125766 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
81.9/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Il2
- Entrez Gene ID
-
16183
- Gene Name
- interleukin 2
- Gene Aliases
-
Il-2
- Chromosome Location
-
Chr.3: 37120713 - 37125954 on GRCm38
- UniGene ID
- Mm.14190
- Species
-
Mouse