gRNA Design Details
- Target DNA Sequence
- TTGGCCGCGGATTTTCGACA
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 85376371 - 85376393 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sh3gl2
- Entrez Gene ID
-
20404
- Gene Name
- SH3-domain GRB2-like 2
- Gene Aliases
-
9530001L19Rik, AI120490, AW555077, B930049H17Rik, EEN-B1, SH3PA, Sh3d2a
- Chromosome Location
-
Chr.4: 85205456 - 85389380 on GRCm38
- UniGene ID
- Mm.143603
- Species
-
Mouse