gRNA Design Details
- Target DNA Sequence
- GTCCCGGTACGAGCACTTGT
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 75972290 - 75972312 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Kdr
- Entrez Gene ID
-
16542
- Gene Name
- kinase insert domain protein receptor
- Gene Aliases
-
6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2
- Chromosome Location
-
Chr.5: 75932827 - 75979074 on GRCm38
- UniGene ID
- Mm.285
- Species
-
Mouse