gRNA Design Details
- Target DNA Sequence
- AACACTCACCATTCCCAGGG
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 75972331 - 75972353 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.51/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Kdr
- Entrez Gene ID
-
16542
- Gene Name
- kinase insert domain protein receptor
- Gene Aliases
-
6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2
- Chromosome Location
-
Chr.5: 75932827 - 75979074 on GRCm38
- UniGene ID
- Mm.285
- Species
-
Mouse