gRNA Design Details
- Target DNA Sequence
- TGGAACACTTGTAGTGGCCT
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 101080185 - 101080207 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ptgfrn
- Entrez Gene ID
-
19221
- Gene Name
- prostaglandin F2 receptor negative regulator
- Gene Aliases
-
4833445A08Rik, AU042434, CD9P-1, FPRP, Trim45
- Chromosome Location
-
Chr.3: 101040230 - 101111105 on GRCm38
- UniGene ID
- Mm.24807
- Species
-
Mouse