gRNA Design Details
- Target DNA Sequence
- TGTATCAGCTTCATCTACGG
- PAM Sequence
- CGG
- Target locus
-
Chr.7: 29258486 - 29258508 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.59/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spint2
- Entrez Gene ID
-
20733
- Gene Name
- serine protease inhibitor, Kunitz type 2
- Gene Aliases
-
AL024025, C76321, HAI-2
- Chromosome Location
-
Chr.7: 29256330 - 29282064 on GRCm38
- UniGene ID
- Mm.295230
- Species
-
Mouse