gRNA Design Details
- Target DNA Sequence
- TGGCTCATTCATAGGCTATA
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 177131025 - 177131047 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.43/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Akt3
- Entrez Gene ID
-
23797
- Gene Name
- thymoma viral proto-oncogene 3
- Gene Aliases
-
AI851531, D930002M15Rik, Nmf350
- Chromosome Location
-
Chr.1: 177020076 - 177258289 on GRCm38
- UniGene ID
- Mm.235194
- Species
-
Mouse