gRNA Design Details
- Target DNA Sequence
- GACATTCCTCGGCTCTTCCG
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 95020942 - 95020964 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.34/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgrg3
- Entrez Gene ID
-
54672
- Gene Name
- adhesion G protein-coupled receptor G3
- Gene Aliases
-
A030001G24Rik, AW214383, Gpr97, Pb1, Pb99
- Chromosome Location
-
Chr.8: 95017114 - 95045250 on GRCm38
- UniGene ID
- Mm.27995
- Species
-
Mouse