gRNA Design Details
- Target DNA Sequence
- GTCGTATTCACAAAGATAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 23513841 - 23513863 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.75/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Masp1
- Entrez Gene ID
-
17174
- Gene Name
- mannan-binding lectin serine peptidase 1
- Gene Aliases
-
AW048060, CCPII, Crarf, Masp1/3
- Chromosome Location
-
Chr.16: 23451785 - 23521073 on GRCm38
- UniGene ID
- Mm.1213
- Species
-
Mouse