gRNA Design Details
- Target DNA Sequence
- CGAGCTGAGTTTGGCTCGTT
- PAM Sequence
- TGG
- Target locus
-
Chr.16: 4213167 - 4213189 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
83.85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Crebbp
- Entrez Gene ID
-
12914
- Gene Name
- CREB binding protein
- Gene Aliases
-
AW558298, CBP, CBP/p300, KAT3A, p300/CBP
- Chromosome Location
-
Chr.16: 4081334 - 4213454 on GRCm38
- UniGene ID
- Mm.132238
- Species
-
Mouse