gRNA Design Details
- Target DNA Sequence
- GGTATATGAAGCGGCCACCG
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 147376992 - 147377014 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Flt3
- Entrez Gene ID
-
14255
- Gene Name
- FMS-like tyrosine kinase 3
- Gene Aliases
-
B230315G04, CD135, Flk-2, Flk2, Flt-3, Ly72, wmfl
- Chromosome Location
-
Chr.5: 147330741 - 147400489 on GRCm38
- UniGene ID
- Mm.194
- Species
-
Mouse