gRNA Design Details

Target DNA Sequence
GGTTACTCTCCATGTCAGGG
PAM Sequence
AGG
Target locus
Chr.11: 64286515 - 64286537 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.54/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GPR137
Entrez Gene ID
56834
Gene Name
G protein-coupled receptor 137
Gene Aliases
C11orf4, GPR137A, TM7SF1L1
Chromosome Location
Chr.11: 64270337 - 64289500 on GRCh38
UniGene ID
Hs.523763
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001170726.1 NP_001164197.1 3
NM_001170880.1 NP_001164351.1 1
NM_001170881.1 NP_001164352.1 2
NM_001177358.1 NP_001170829.1 2
NM_020155.3 NP_064540.3 1
XM_005274100.2 XP_005274157.1 2
XM_005274101.2 XP_005274158.1 2
XM_005274102.2 XP_005274159.1 2
XM_005274104.2 XP_005274161.1 2
XM_011545168.2 XP_011543470.1 2
XM_011545169.1 XP_011543471.1 2
XM_011545170.2 XP_011543472.1 2
XM_011545171.2 XP_011543473.1 2
XM_011545172.2 XP_011543474.1 2
XM_017018014.1 XP_016873503.1 2
XM_017018015.1 XP_016873504.1 2
XM_017018016.1 XP_016873505.1 2