gRNA Design Details
- Target DNA Sequence
- GGTCGTATTCGTCCACAAAA
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 534223 - 534245 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.77/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HRAS
- Entrez Gene ID
-
3265
- Gene Name
- HRas proto-oncogene, GTPase
- Gene Aliases
-
C-BAS/HAS, C-H-RAS, C-HA-RAS1, CTLO, H-RASIDX, HAMSV, HRAS1, RASH1, p21ras
- Chromosome Location
-
Chr.11: 532242 - 535567 on GRCh38
- UniGene ID
- Hs.37003
- Species
-
Human