gRNA Design Details
- Target DNA Sequence
- GTAGCAGAGTTCAAGTCTTC
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 34358119 - 34358141 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.21/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CCL13
- Entrez Gene ID
-
6357
- Gene Name
- C-C motif chemokine ligand 13
- Gene Aliases
-
CKb10, MCP-4, NCC-1, NCC1, SCYA13, SCYL1
- Chromosome Location
-
Chr.17: 34356452 - 34358610 on GRCh38
- UniGene ID
- Hs.414629
- Species
-
Human