gRNA Design Details

Target DNA Sequence
AAGAGATAGCGCAGTTGGCG
PAM Sequence
CGG
Target locus
Chr.9: 34637557 - 34637579 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 97.27/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SIGMAR1
Entrez Gene ID
10280
Gene Name
sigma non-opioid intracellular receptor 1
Gene Aliases
ALS16, DSMA2, OPRS1, SIG-1R, SR-BP, SR-BP1, SRBP, hSigmaR1, sigma1R
Chromosome Location
Chr.9: 34634722 - 34637826 on GRCh38
UniGene ID
Hs.522087
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001282205.1 NP_001269134.1 1
NM_001282206.1 NP_001269135.1 1
NM_001282208.1 NP_001269137.1 1
NM_001282209.1 NP_001269138.1 1
NM_005866.3 NP_005857.1 1
NM_147157.2 NP_671513.1 1
NR_104108.1 1