gRNA Design Details

Target DNA Sequence
TTCGGTGATCTTTGGTCTGA
PAM Sequence
AGG
Target locus
Chr.2: 238188231 - 238188253 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 92.184/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ILKAP
Entrez Gene ID
80895
Gene Name
ILK associated serine/threonine phosphatase
Gene Aliases
ILKAP2, ILKAP3, PP2C-DELTA, PPM1O
Chromosome Location
Chr.2: 238170401 - 238203702 on GRCh38
UniGene ID
Hs.92033
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_030768.2 NP_110395.1 5
XM_005246106.1 XP_005246163.1 5
XM_006712784.1 XP_006712847.1 5
XM_017005056.1 XP_016860545.1 5
XM_017005057.1 XP_016860546.1 5