gRNA Design Details

Target DNA Sequence
GCCATTCTTCTCTTCCTCGG
PAM Sequence
AGG
Target locus
Chr.2: 238189887 - 238189909 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.414/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ILKAP
Entrez Gene ID
80895
Gene Name
ILK associated serine/threonine phosphatase
Gene Aliases
ILKAP2, ILKAP3, PP2C-DELTA, PPM1O
Chromosome Location
Chr.2: 238170401 - 238203702 on GRCh38
UniGene ID
Hs.92033
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_030768.2 NP_110395.1 4
XM_005246106.1 XP_005246163.1 4
XM_006712784.1 XP_006712847.1 4
XM_011511946.2 XP_011510248.1 4
XM_017005056.1 XP_016860545.1 4
XM_017005057.1 XP_016860546.1 4
XM_017005058.1 XP_016860547.1 4