gRNA Design Details

Target DNA Sequence
TAACGGTGCCGTTAGGCAGA
PAM Sequence
AGG
Target locus
Chr.8: 37797508 - 37797530 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 95.64/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADGRA2
Entrez Gene ID
25960
Gene Name
adhesion G protein-coupled receptor A2
Gene Aliases
GPR124, TEM5
Chromosome Location
Chr.8: 37796883 - 37843986 on GRCh38
UniGene ID
Hs.274136
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_032777.9 NP_116166.9 1