gRNA Design Details
- Target DNA Sequence
- GAATATGATGAGGACGACCT
- PAM Sequence
- GGG
- Target locus
-
Chr.17: 36952948 - 36952970 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AATF
- Entrez Gene ID
-
26574
- Gene Name
- apoptosis antagonizing transcription factor
- Gene Aliases
-
BFR2, CHE-1, CHE1, DED
- Chromosome Location
-
Chr.17: 36948875 - 37056871 on GRCh38
- UniGene ID
- Hs.195740
- Species
-
Human