gRNA Design Details

Target DNA Sequence
CCTCCGGAGAAAAGCGGGTT
PAM Sequence
TGG
Target locus
Chr.4: 56821553 - 56821575 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 88.01/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SPINK2
Entrez Gene ID
6691
Gene Name
serine peptidase inhibitor, Kazal type 2
Gene Aliases
HUSI-II
Chromosome Location
Chr.4: 56809860 - 56822210 on GRCh38
UniGene ID
Hs.98243
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001271718.1 NP_001258647.1 1
NM_001271719.1 NP_001258648.1 1
NM_001271720.1 NP_001258649.1 1
NM_001271721.1 NP_001258650.1 1
NM_001271722.1 NP_001258651.1 1