gRNA Design Details

Target DNA Sequence
CCAGATTGATTGGGCTGCAT
PAM Sequence
TGG
Target locus
Chr.6: 99412716 - 99412738 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 87.29/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PNISR
Entrez Gene ID
25957
Gene Name
PNN interacting serine and arginine rich protein
Gene Aliases
C6orf111, HSPC306, SFRS18, SRrp130, bA98I9.2
Chromosome Location
Chr.6: 99398051 - 99425345 on GRCh38
UniGene ID
Hs.520287
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001322405.1 NP_001309334.1 4
NM_001322406.1 NP_001309335.1 5
NM_001322408.1 NP_001309337.1 3
NM_001322410.1 NP_001309339.1 4
NM_001322412.1 NP_001309341.1 4
NM_001322413.1 NP_001309342.1 3
NM_001322414.1 NP_001309343.1 4
NM_001322415.1 NP_001309344.1 3
NM_001322416.1 NP_001309345.1 4
NM_001322417.1 NP_001309346.1 4
NM_001322418.1 NP_001309347.1 3
NM_001322419.1 NP_001309348.1 4
NM_015491.2 NP_056306.1 3
NM_032870.3 NP_116259.2 4
NR_136326.1 3
XM_005266912.3 XP_005266969.2 3
XM_017010710.1 XP_016866199.1 4
XM_017010711.1 XP_016866200.1 3
XM_017010712.1 XP_016866201.1 4
XM_017010713.1 XP_016866202.1 4