gRNA Design Details
- Target DNA Sequence
- GATTCAAGAGAGCCCAGAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 142257674 - 142257696 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.67/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS58
- Entrez Gene ID
-
136541
- Gene Name
- protease, serine 58
- Gene Aliases
-
PRSS1, TRY1, TRYX3, UNQ2540
- Chromosome Location
-
Chr.7: 142252138 - 142258058 on GRCh38
- UniGene ID
- Hs.98609
- Species
-
Human