gRNA Design Details
- Target DNA Sequence
- TCACGCTTCTCTATTACGGC
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 130971512 - 130971534 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
95.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRD1
- Entrez Gene ID
-
283383
- Gene Name
- adhesion G protein-coupled receptor D1
- Gene Aliases
-
GPR133, PGR25
- Chromosome Location
-
Chr.12: 130953907 - 131141469 on GRCh38
- UniGene ID
- Hs.719239
- Species
-
Human