gRNA Design Details

Target DNA Sequence
GACCAATCCCTTCTGCGTAT
PAM Sequence
GGG
Target locus
Chr.12: 130981929 - 130981951 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 91.23/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADGRD1
Entrez Gene ID
283383
Gene Name
adhesion G protein-coupled receptor D1
Gene Aliases
GPR133, PGR25
Chromosome Location
Chr.12: 130953907 - 131141469 on GRCh38
UniGene ID
Hs.719239
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_198827.3 NP_942122.2 5