gRNA Design Details

Target DNA Sequence
TATAGGCCCTGGGCAACTGC
PAM Sequence
AGG
Target locus
Chr.16: 31086780 - 31086802 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 86.81/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRSS53
Entrez Gene ID
339105
Gene Name
protease, serine 53
Gene Aliases
POL3S, UNQ308
Chromosome Location
Chr.16: 31083424 - 31090035 on GRCh38
UniGene ID
Hs.569575
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001039503.2 NP_001034592.1 4
XM_011545816.1 XP_011544118.1 4
XM_011545817.1 XP_011544119.1 4
XM_011545818.2 XP_011544120.1 4
XM_011545819.1 XP_011544121.1 4
XM_011545820.1 XP_011544122.1 4