gRNA Design Details
- Target DNA Sequence
- CTTCATCACCCAGCTGAGAG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 30968863 - 30968885 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
83.93/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GHRHR
- Entrez Gene ID
-
2692
- Gene Name
- growth hormone releasing hormone receptor
- Gene Aliases
-
GHRFR, GRFR, IGHD1B
- Chromosome Location
-
Chr.7: 30964021 - 30979531 on GRCh38
- UniGene ID
- Hs.733003
- Species
-
Human