gRNA Design Details

Target DNA Sequence
TCTCACCTATGCTGCCAAAT
PAM Sequence
TGG
Target locus
Chr.17: 2317971 - 2317993 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 89.29/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SRR
Entrez Gene ID
63826
Gene Name
serine racemase
Gene Aliases
ILV1, ISO1
Chromosome Location
Chr.17: 2303371 - 2325267 on GRCh38
UniGene ID
Hs.461954
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001304803.1 NP_001291732.1 2
NM_021947.2 NP_068766.1 3
XM_006721565.3 XP_006721628.1 3
XM_006721566.3 XP_006721629.1 3
XM_011523974.2 XP_011522276.1 3