gRNA Design Details
- Target DNA Sequence
- AGCTTGTTAGCACTGGTGTG
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 2315632 - 2315654 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRR
- Entrez Gene ID
-
63826
- Gene Name
- serine racemase
- Gene Aliases
-
ILV1, ISO1
- Chromosome Location
-
Chr.17: 2303371 - 2325267 on GRCh38
- UniGene ID
- Hs.461954
- Species
-
Human