gRNA Design Details

Target DNA Sequence
TATTGTAGGGTGGTTCGTGT
PAM Sequence
GGG
Target locus
Chr.1: 66635952 - 66635974 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.58/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SGIP1
Entrez Gene ID
84251
Gene Name
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 66533272 - 66751139 on GRCh38
UniGene ID
Hs.132121
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_032291.3 NP_115667.2 4
XM_006710967.2 XP_006711030.1 4
XM_011542291.1 XP_011540593.1 4
XM_011542292.1 XP_011540594.1 4
XM_017002505.1 XP_016857994.1 4
XM_017002506.1 XP_016857995.1 5
XM_017002507.1 XP_016857996.1 4
XM_017002508.1 XP_016857997.1 5
XM_017002509.1 XP_016857998.1 4
XM_017002510.1 XP_016857999.1 5
XM_017002511.1 XP_016858000.1 5
XM_017002512.1 XP_016858001.1 5
XM_017002513.1 XP_016858002.1 4
XM_017002517.1 XP_016858006.1 5
XM_017002524.1 XP_016858013.1 5
XM_017002525.1 XP_016858014.1 5
XM_017002526.1 XP_016858015.1 4
XM_017002527.1 XP_016858016.1 5
XM_017002530.1 XP_016858019.1 4
XM_017002533.1 XP_016858022.1 5