gRNA Design Details
- Target DNA Sequence
- AGGCTACAGCATCAGACCCG
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 66642833 - 66642855 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.63/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SGIP1
- Entrez Gene ID
-
84251
- Gene Name
- SH3 domain GRB2 like endophilin interacting protein 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 66533272 - 66751139 on GRCh38
- UniGene ID
- Hs.132121
- Species
-
Human